Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036629.2 | NP_001031706.1 | 10 - 11 | 1364 | 80 | |
NM_118530.5 | NP_194129.1 | 10 - 11 | 1419 | 80 | ||
GenBank mRNA | AF082176.1 | - | 10 - 11 | 1257 | 80 | |
AF344312.1 | - | 10 - 11 | 970 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |