Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118536.4 | NP_194135.1 | 5 - 6 | 1365 | 81 | |
GenBank mRNA | AK118407.1 | - | 5 - 6 | 1222 | 81 | |
BT010732.1 | - | 5 - 6 | 1163 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |