Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIPK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084973.1 | NP_001078442.1 | 7 - 8 | 783 | 148 | |
NM_118573.4 | NP_194171.1 | 7 - 8 | 1060 | 148 | ||
GenBank mRNA | AF290193.1 | - | 7 - 8 | 660 | 148 | |
BT000958.1 | - | 7 - 8 | 660 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |