Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PWD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341673.1 | NP_001328878.1 | 7 - 8 | 860 | 68 | |
NM_001341674.1 | NP_001328879.1 | 7 - 8 | 864 | 68 | ||
NM_118578.5 | NP_194176.3 | 6 - 7 | 662 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |