Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203892.1 | NP_001190821.1 | 1 - 2 | 532 | 76 | |
NM_118580.4 | NP_194178.1 | 1 - 2 | 552 | 76 | ||
NM_179104.3 | NP_849435.1 | 2 - 3 | 434 | 76 | ||
GenBank mRNA | AY086038.1 | - | 1 - 2 | 450 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |