Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118595.5 | NP_194193.2 | 13 - 14 | 1920 | 76 | |
GenBank mRNA | AK227111.1 | - | 13 - 14 | 1758 | 76 | |
BT000953.1 | - | 13 - 14 | 1705 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |