Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084975.1 | NP_001078444.1 | 1 - 2 | 739 | 67 | |
NM_118595.5 | NP_194193.2 | 1 - 2 | 739 | 67 | ||
GenBank mRNA | AK227111.1 | - | 1 - 2 | 577 | 67 | |
BT000953.1 | - | 1 - 2 | 524 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |