Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084975.1 | NP_001078444.1 | 9 - 10 | 1482 | 108 | |
NM_118595.5 | NP_194193.2 | 9 - 10 | 1482 | 108 | ||
GenBank mRNA | AK227111.1 | - | 9 - 10 | 1320 | 108 | |
BT000953.1 | - | 9 - 10 | 1267 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |