Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341698.1 | NP_001329102.1 | 4 - 5 | 508 | 137 | |
NM_001341699.1 | NP_001329107.1 | 7 - 8 | 908 | 137 | ||
NM_001341700.1 | NP_001329103.1 | 7 - 8 | 908 | 137 | ||
NM_001341701.1 | NP_001329104.1 | 5 - 6 | 1659 | 137 | ||
NM_001341702.1 | NP_001329105.1 | 5 - 6 | 1659 | 137 | ||
NM_001341703.1 | NP_001329106.1 | 6 - 7 | 1514 | 137 | ||
NM_001341704.1 | NP_001329108.1 | 3 - 4 | 1495 | 137 | ||
NM_001341705.1 | NP_001329109.1 | 6 - 7 | 1042 | 137 | ||
NM_118607.3 | NP_194205.1 | 7 - 8 | 942 | 137 | ||
NM_202881.2 | NP_974610.1 | 6 - 7 | 752 | 137 | ||
GenBank mRNA | AK227055.1 | - | 7 - 8 | 814 | 137 | |
BT025991.1 | - | 7 - 8 | 729 | 137 | ||
D45353.1 | - | 7 - 8 | 849 | 137 | ||
U16177.1 | - | 7 - 8 | 772 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |