Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAE1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118626.3 | NP_567712.1 | 3 - 4 | 483 | 109 | |
GenBank mRNA | AF510523.1 | - | 3 - 4 | 345 | 109 | |
AY072375.1 | - | 3 - 4 | 482 | 109 | ||
AY086568.1 | - | 3 - 4 | 482 | 109 | ||
BT000094.1 | - | 3 - 4 | 328 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |