Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203902.1 | NP_001190831.1 | 1 - 2 | 229 | 74 | |
NM_118637.4 | NP_194235.1 | 1 - 2 | 227 | 74 | ||
GenBank mRNA | AY039527.1 | - | 1 - 2 | 120 | 74 | |
AY050471.1 | - | 1 - 2 | 57 | 74 | ||
AY084704.1 | - | 1 - 2 | 168 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |