Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036634.1 | NP_001031711.1 | 1 - 2 | 78 | ||
NM_118643.5 | NP_194241.3 | 1 - 2 | 788 | 81 | ||
GenBank mRNA | AK175293.1 | - | 1 - 2 | 78 | ||
AK221790.1 | - | 1 - 2 | 788 | 81 | ||
AY219827.1 | - | 1 - 2 | 479 | 81 | ||
AY322526.1 | - | 1 - 2 | 479 | 81 | ||
BT026456.1 | - | 1 - 2 | 479 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |