Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036634.1 | NP_001031711.1 | 3 - 4 | 1240 | 96 | |
NM_118643.5 | NP_194241.3 | 3 - 4 | 1244 | 96 | ||
GenBank mRNA | AK175293.1 | - | 3 - 4 | 1051 | 96 | |
AK221790.1 | - | 3 - 4 | 1244 | 96 | ||
AY219827.1 | - | 3 - 4 | 935 | 96 | ||
AY322526.1 | - | 3 - 4 | 935 | 96 | ||
BT026456.1 | - | 3 - 4 | 935 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |