Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341722.1 | NP_001320059.1 | 21 - 22 | 2921 | 83 | |
GenBank mRNA | GQ148553.1 | - | 21 - 22 | 2828 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |