Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GA20OX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341731.1 | NP_001328762.1 | 1 - 2 | 728 | 96 | |
NM_118674.5 | NP_194272.1 | 1 - 2 | 728 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |