Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118679.2 | NP_567719.1 | 1 | 891 | 160 | |
GenBank mRNA | AB007789.1 | - | 1 | 876 | 160 | |
AF074601.1 | - | 1 | 749 | 160 | ||
EF523038.1 | - | 1 | 876 | 160 | ||
EF523040.1 | - | 1 | 876 | 160 | ||
EF523053.1 | - | 1 | 876 | 160 | ||
EF523056.1 | - | 1 | 876 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |