Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118681.4 | NP_567721.1 | 1 | 1013 | 69 | |
GenBank mRNA | AB007788.1 | - | 1 | 742 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Inventoried Both primers and probe map within a single exon |