Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACYB-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341751.1 | NP_001328918.1 | 2 - 3 | 474 | 72 | |
NM_118689.4 | NP_567723.1 | 2 - 3 | 300 | 72 | ||
GenBank mRNA | AY120730.1 | - | 2 - 3 | 296 | 72 | |
BT002176.1 | - | 2 - 3 | 192 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |