Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341753.1 | NP_001329121.1 | 1 - 2 | 862 | 80 | |
NM_118691.3 | NP_194289.2 | 2 - 3 | 462 | 80 | ||
GenBank mRNA | AK176093.1 | - | 2 - 3 | 319 | 80 | |
BT010513.1 | - | 1 - 2 | 241 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |