Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACD1-LIKE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118697.4 | NP_567725.1 | 5 - 6 | 1273 | 65 | |
NM_179113.2 | NP_849444.1 | 4 - 5 | 1279 | 65 | ||
GenBank mRNA | AF424575.1 | - | 5 - 6 | 1179 | 65 | |
AY056168.1 | - | 5 - 6 | 1202 | 65 | ||
AY062731.1 | - | 4 - 5 | 1279 | 65 | ||
AY344062.1 | - | 5 - 6 | 1166 | 65 | ||
BT002586.1 | - | 4 - 5 | 1235 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |