Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFU3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118724.4 | NP_567735.1 | 3 - 4 | 718 | 77 | |
GenBank mRNA | AJ512935.1 | - | 3 - 4 | 544 | 77 | |
AK317114.1 | - | 3 - 4 | 664 | 77 | ||
BT024624.1 | - | 3 - 4 | 519 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |