Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAP1;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118744.3 | NP_194341.1 | 11 - 12 | 1194 | 85 | |
GenBank mRNA | AY054484.1 | - | 11 - 12 | 1111 | 85 | |
AY093283.1 | - | 11 - 12 | 1050 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |