Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIOX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203915.2 | NP_001190844.1 | 2 - 3 | 311 | 67 | |
NM_118759.5 | NP_194356.2 | 2 - 3 | 416 | 67 | ||
GenBank mRNA | AY142501.1 | - | 2 - 3 | 172 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |