Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNE17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341795.1 | NP_001328493.1 | 1 - 2 | 142 | 93 | |
NM_118766.2 | NP_567744.1 | 1 - 2 | 142 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |