Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNE17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341795.1 | NP_001328493.1 | 7 - 8 | 1616 | 84 | |
NM_118766.2 | NP_567744.1 | 7 - 8 | 1658 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |