Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSN6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118776.3 | NP_567746.1 | 2 - 3 | 627 | 101 | |
GenBank mRNA | AF395064.1 | - | 2 - 3 | 390 | 101 | |
AY087556.1 | - | 2 - 3 | 397 | 101 | ||
BT002345.1 | - | 2 - 3 | 390 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |