Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118777.2 | NP_194374.1 | 2 - 3 | 1012 | 77 | |
GenBank mRNA | AY052649.1 | - | 2 - 3 | 1011 | 77 | |
BT030361.1 | - | 2 - 3 | 1011 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |