Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160797.2 | NP_001154269.1 | 1 - 2 | 1337 | 74 | |
GenBank mRNA | AK118341.1 | - | 1 - 2 | 1252 | 74 | |
AK228147.1 | - | 1 - 2 | 1175 | 74 | ||
BT003145.1 | - | 1 - 2 | 883 | 74 | ||
EF554923.1 | - | 1 - 2 | 883 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |