Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT-HF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118824.4 | NP_194420.1 | 12 - 13 | 1241 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |