Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATRBP45C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118834.4 | NP_567764.1 | 3 - 4 | 983 | 91 | |
GenBank mRNA | AF370339.1 | - | 3 - 4 | 922 | 91 | |
AY054487.1 | - | 3 - 4 | 914 | 91 | ||
AY062999.1 | - | 3 - 4 | 831 | 91 | ||
AY093292.1 | - | 3 - 4 | 831 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |