Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDIL5-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118842.4 | NP_567765.2 | 3 - 4 | 439 | 156 | |
NM_148376.2 | NP_680742.2 | 4 - 5 | 471 | 156 | ||
GenBank mRNA | AY054496.1 | - | 3 - 4 | 314 | 156 | |
AY093266.1 | - | 3 - 4 | 231 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |