Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341838.1 | NP_001328096.1 | 9 - 10 | 1435 | 142 | |
NM_118852.3 | NP_567768.1 | 10 - 11 | 1618 | 142 | ||
GenBank mRNA | D21137.1 | - | 10 - 11 | 1462 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |