Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203925.2 | NP_001190854.1 | 2 - 3 | 444 | 103 | |
NM_001341872.1 | NP_001320077.1 | 2 - 3 | 467 | 103 | ||
NM_001341873.1 | NP_001329564.1 | 2 - 3 | 435 | 103 | ||
NM_001341874.1 | NP_001329563.1 | 2 - 3 | 467 | 103 | ||
NM_001341875.1 | NP_001329565.1 | 2 - 3 | 435 | 103 | ||
NM_118899.4 | NP_194493.2 | 2 - 3 | 593 | 103 | ||
GenBank mRNA | HM776217.1 | - | 2 - 3 | 321 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Arabidopsis thaliana Probe spans exons |