Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLE13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148652.3 | NP_683493.1 | 1 | 472 | 58 | |
GenBank mRNA | AK220675.1 | - | 1 | 302 | 58 | |
BT011710.1 | - | 1 | 277 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |