Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334857.1 | NP_001321745.1 | 9 - 10 | 1649 | 82 | |
NM_001334858.1 | NP_001321744.1 | 9 - 10 | 1649 | 82 | ||
NM_001334859.1 | NP_001321746.1 | 9 - 10 | 1649 | 82 | ||
NM_001334860.1 | NP_001321747.1 | 9 - 10 | 1642 | 82 | ||
NM_148667.2 | NP_683508.1 | 10 - 11 | 1482 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |