Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334857.1 | NP_001321745.1 | 8 - 9 | 1518 | 103 | |
NM_001334858.1 | NP_001321744.1 | 8 - 9 | 1518 | 103 | ||
NM_001334859.1 | NP_001321746.1 | 8 - 9 | 1518 | 103 | ||
NM_001334860.1 | NP_001321747.1 | 8 - 9 | 1511 | 103 | ||
NM_148667.2 | NP_683508.1 | 9 - 10 | 1351 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |