Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLA18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112002.3 | NP_566398.1 | 2 - 3 | 1792 | 118 | |
GenBank mRNA | AY056197.1 | - | 2 - 3 | 1604 | 118 | |
AY058879.1 | - | 2 - 3 | 1611 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |