Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATKRS-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112003.4 | NP_187777.1 | 11 - 12 | 1546 | 74 | |
GenBank mRNA | AF125574.1 | - | 11 - 12 | 1529 | 74 | |
AY099551.1 | - | 11 - 12 | 1507 | 74 | ||
BT002123.1 | - | 11 - 12 | 1443 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |