Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202935.1 | NP_001189864.1 | 17 - 18 | 2293 | 128 | |
NM_001337961.1 | NP_001319526.1 | 17 - 18 | 2293 | 128 | ||
NM_001337962.1 | NP_001326292.1 | 17 - 18 | 2296 | 128 | ||
NM_112024.6 | NP_187797.3 | 17 - 18 | 2222 | 128 | ||
GenBank mRNA | BT002760.1 | - | 17 - 18 | 2023 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |