Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084669.2 | NP_001078138.1 | 6 - 7 | 967 | 89 | |
NM_001161137.1 | NP_001154609.1 | 7 - 8 | 961 | 89 | ||
GenBank mRNA | DQ231060.1 | - | 6 - 7 | 866 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |