Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSB28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119008.4 | NP_567814.1 | 1 - 2 | 231 | 75 | |
GenBank mRNA | AF462867.1 | - | 1 - 2 | 85 | 75 | |
AY086344.1 | - | 1 - 2 | 164 | 75 | ||
AY113018.1 | - | 1 - 2 | 72 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |