Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAK1AT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341955.1 | NP_001329562.1 | 1 - 2 | 533 | 104 | |
NM_119042.5 | NP_194627.1 | 1 - 2 | 575 | 104 | ||
NM_179137.3 | NP_849468.1 | 1 - 2 | 593 | 104 | ||
GenBank mRNA | AB009399.1 | - | 1 - 2 | 527 | 104 | |
AK316966.1 | - | 1 - 2 | 550 | 104 | ||
AY042816.1 | - | 1 - 2 | 524 | 104 | ||
AY064627.1 | - | 1 - 2 | 501 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |