Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341965.1 | NP_001328427.1 | 5 - 6 | 723 | 66 | |
NM_119078.5 | NP_194662.2 | 5 - 6 | 831 | 66 | ||
GenBank mRNA | AY063817.1 | - | 5 - 6 | 564 | 66 | |
AY096457.1 | - | 5 - 6 | 447 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |