Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFIM-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119128.3 | NP_567835.1 | 1 - 2 | 386 | 95 | |
GenBank mRNA | AY048229.1 | - | 1 - 2 | 349 | 95 | |
AY085984.1 | - | 1 - 2 | 351 | 95 | ||
AY091704.1 | - | 1 - 2 | 187 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |