Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFIM-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119128.3 | NP_567835.1 | 5 - 6 | 682 | 105 | |
GenBank mRNA | AY048229.1 | - | 5 - 6 | 645 | 105 | |
AY085984.1 | - | 5 - 6 | 647 | 105 | ||
AY091704.1 | - | 5 - 6 | 483 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |