Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341980.1 | NP_001327988.1 | 29 - 30 | 3365 | 108 | |
NM_119136.4 | NP_194719.1 | 29 - 30 | 3055 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |