Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341980.1 | NP_001327988.1 | 33 - 34 | 3741 | 71 | |
NM_119136.4 | NP_194719.1 | 33 - 34 | 3431 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |