Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VLN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203940.1 | NP_001190869.1 | 8 - 9 | 1036 | 106 | |
NM_001341999.1 | NP_001329918.1 | 8 - 9 | 1104 | 106 | ||
NM_001342000.1 | NP_001329917.1 | 7 - 8 | 1034 | 106 | ||
NM_119162.5 | NP_194745.1 | 7 - 8 | 1376 | 106 | ||
GenBank mRNA | AK117296.1 | - | 7 - 8 | 983 | 106 | |
BT005980.1 | - | 6 - 7 | 773 | 106 | ||
Y12782.1 | - | 1016 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |