Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUXF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203943.1 | NP_001190872.1 | 3 - 4 | 258 | 76 | |
NM_119168.4 | NP_194751.1 | 3 - 4 | 337 | 76 | ||
GenBank mRNA | AK227757.1 | - | 3 - 4 | 168 | 76 | |
AY087616.1 | - | 3 - 4 | 199 | 76 | ||
BT006382.1 | - | 2 - 3 | 135 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |