Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119182.4 | NP_194765.2 | 3 - 4 | 1318 | 94 | |
GenBank mRNA | AK227176.1 | - | 3 - 4 | 1295 | 94 | |
AY124000.1 | - | 3 - 4 | 1294 | 94 | ||
BT002284.1 | - | 3 - 4 | 1078 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |